ENA447956907 Enamine

3-[1-(2,2,2-trifluoroethyl)-1H-1,2,4-triazol-3-yl]morpholine hydrochloride

  • MDL number MFCD28383929
  • Cas Number : 1803587-62-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205678

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205678
Availability details for
Quantity
  • Availability
  • Add to Cart