ENA447956916 Enamine

4,4-dimethyl-1-(1,3-thiazol-4-yl)pentan-1-amine dihydrochloride

  • MDL number MFCD28383937
  • Cas Number : 1803562-17-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205787

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205787
Availability details for
Quantity
  • Availability
  • Add to Cart