ENA447956918 Enamine

diethyl [(2,6-dimethylphenyl)methyl]phosphonate

  • MDL number MFCD28383940
  • Cas Number : 902526-55-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205836

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205836
Availability details for
Quantity
  • Availability
  • Add to Cart