ENA447956931 Enamine

5-amino-1H-1,2,3-triazole-4-carboximidamide dihydrochloride

  • MDL number MFCD28383954
  • Cas Number : 1803604-51-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206119

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206119
Availability details for
Quantity
  • Availability
  • Add to Cart