ENA447956934 Enamine

2-chloro-N-methylbenzene-1-carboximidamide hydrochloride

  • MDL number MFCD28383959
  • Cas Number : 1803598-95-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206339

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206339
Availability details for
Quantity
  • Availability
  • Add to Cart