ENA447956935 Enamine

methyl 2-(oxolan-3-yl)ethanecarboximidate hydrochloride

  • MDL number MFCD28383960
  • Cas Number : 1803584-59-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206340

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206340
Availability details for
Quantity
  • Availability
  • Add to Cart