ENA447956936 Enamine

2-(4-chlorophenyl)-2-(cyclopropylmethoxy)ethan-1-amine hydrochloride

  • MDL number MFCD28383961
  • Cas Number : 1803570-57-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206341

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206341
Availability details for
Quantity
  • Availability
  • Add to Cart