ENA447956937 Enamine

1-(2-aminophenyl)-2-methylprop-2-en-1-one; trifluoroacetic acid

  • MDL number MFCD28383962
  • Cas Number : 1803605-37-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data CF3COOH
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206357

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206357
Availability details for
Quantity
  • Availability
  • Add to Cart