ENA447956941 Enamine

{[(2S,4S)-4-fluoro-1-[(5-methyl-1,2,4-oxadiazol-3-yl)methyl]pyrrolidin-2-yl]methyl}(methyl)amine dihydrochloride

  • MDL number MFCD28383965
  • Cas Number : 1807941-78-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206401

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206401
Availability details for
Quantity
  • Availability
  • Add to Cart