ENA447956947 Enamine

tert-butyl N-(4-{[N'-(methoxycarbonyl)hydrazinecarbonyl]amino}phenyl)carbamate

  • MDL number MFCD28397419
  • Cas Number : 1803562-66-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206437

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206437
Availability details for
Quantity
  • Availability
  • Add to Cart