ENA447956948 Enamine

4-[4-(methoxymethoxy)phenyl]-1,2,4-triazolidine-3,5-dione

  • MDL number MFCD28397420
  • Cas Number : 1803566-46-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206444

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206444
Availability details for
Quantity
  • Availability
  • Add to Cart