ENA447956949 Enamine

N-({[4-(methoxymethoxy)phenyl]carbamoyl}amino)methoxyformamide

  • MDL number MFCD28397421
  • Cas Number : 1803605-46-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206445

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206445
Availability details for
Quantity
  • Availability
  • Add to Cart