ENA447956953 Enamine

2-(aminooxy)-3-methylbutanoic acid hydrochloride

  • MDL number MFCD28383974
  • Cas Number : 42990-22-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206507

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206507
Availability details for
Quantity
  • Availability
  • Add to Cart