ENA447956958 Enamine

2-(1H-imidazol-2-yl)-2-phenylethan-1-amine dihydrochloride

  • MDL number MFCD28383981
  • Cas Number : 1803605-54-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206538

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206538
Availability details for
Quantity
  • Availability
  • Add to Cart