ENA447956960 Enamine

[(1H-imidazol-2-yl)methyl][2-(2-methylphenyl)ethyl]amine hydrochloride

  • MDL number MFCD28383984
  • Cas Number : 1803604-53-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206567

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206567
Availability details for
Quantity
  • Availability
  • Add to Cart