ENA447956963 Enamine

4,4-dimethyl-1-(1-methyl-1H-imidazol-2-yl)pentan-1-amine hydrochloride

  • MDL number MFCD28383988
  • Cas Number : 1803605-52-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206614

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206614
Availability details for
Quantity
  • Availability
  • Add to Cart