ENA447956964 Enamine

2,2-difluoro-2-(3-methylphenyl)ethan-1-amine hydrochloride

  • MDL number MFCD28383989
  • Cas Number : 1803562-69-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206616

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206616
Availability details for
Quantity
  • Availability
  • Add to Cart