ENA447956970 Enamine

(2R)-1-(ethylamino)propan-2-ol hydrochloride

  • MDL number MFCD28383996
  • Cas Number : 1807914-22-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206692

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206692
Availability details for
Quantity
  • Availability
  • Add to Cart