ENA447956973 Enamine

[1,2,4]triazolo[1,5-a]pyrimidine hydrochloride

  • MDL number MFCD28397425
  • Cas Number : 956074-70-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206730

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206730
Availability details for
Quantity
  • Availability
  • Add to Cart