ENA447956976 Enamine

4-(4-ethylpiperazin-1-yl)-2-methoxyaniline dihydrochloride

  • MDL number MFCD28384000
  • Cas Number : 1803570-62-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206809

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206809
Availability details for
Quantity
  • Availability
  • Add to Cart