ENA447956977 Enamine

methyl 5-chloro-2-oxopentanoate

  • MDL number MFCD28384001
  • Cas Number : 1803596-87-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206821

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206821
Availability details for
Quantity
  • Availability
  • Add to Cart