ENA447956981 Enamine

1-[2-(propan-2-yloxy)phenyl]piperazine dihydrochloride

  • MDL number MFCD28384006
  • Cas Number : 171855-28-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206919

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206919
Availability details for
Quantity
  • Availability
  • Add to Cart