ENA447956989 Enamine

1-(4-amino-6-bromoquinolin-3-yl)ethan-1-one

  • MDL number MFCD28397431
  • Cas Number : 1803605-58-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207080

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207080
Availability details for
Quantity
  • Availability
  • Add to Cart