ENA447956990 Enamine

3-bromo-4-chloro-5-methylpyridine hydrochloride

  • MDL number MFCD28384012
  • Cas Number : 1803562-20-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207090

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207090
Availability details for
Quantity
  • Availability
  • Add to Cart