ENA447956991 Enamine

4-(2-aminoethyl)oxan-4-ol hydrochloride

  • MDL number MFCD20485337
  • Cas Number : 910377-18-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207092

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207092
Availability details for
Quantity
  • Availability
  • Add to Cart