ENA447957005 Enamine

2-{[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}-1lambda6,2-thiazolidine-1,1-dione

  • MDL number MFCD28397437
  • Cas Number : 2246525-96-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207262

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207262
Availability details for
Quantity
  • Availability
  • Add to Cart