ENA447957008 Enamine

4-(benzylsulfanyl)-3-(difluoromethyl)-1H-pyrazole

  • MDL number MFCD28397439
  • Cas Number : 1803562-25-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207314

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207314
Availability details for
Quantity
  • Availability
  • Add to Cart