ENA447957012 Enamine

4-methoxy-4-[3-(2,2,2-trifluoroethyl)-1,2,4-oxadiazol-5-yl]piperidine hydrochloride

  • MDL number MFCD28397442
  • Cas Number : 1803571-03-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207333

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207333
Availability details for
Quantity
  • Availability
  • Add to Cart