ENA447957017 Enamine

5-nitro-6-(1,3-thiazol-5-yl)piperidin-2-one

  • MDL number MFCD28397447
  • Cas Number : 1803571-10-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-207352

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207352
Availability details for
Quantity
  • Availability
  • Add to Cart