ENA447957022 Enamine

N-benzyl-N-cyclopropylazetidin-3-amine hydrochloride

  • MDL number MFCD28384028
  • Cas Number : 1803588-34-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207367

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207367
Availability details for
Quantity
  • Availability
  • Add to Cart