ENA447957025 Enamine

4-(carbonochloridoyl)phenyl ethyl carbonate

  • MDL number MFCD28397451
  • Cas Number : 57045-26-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207401

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207401
Availability details for
Quantity
  • Availability
  • Add to Cart