ENA447957028 Enamine

2-(1H-pyrazol-1-yl)phenol hydrobromide

  • MDL number MFCD28397456
  • Cas Number : 1803562-27-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207458

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207458
Availability details for
Quantity
  • Availability
  • Add to Cart