ENA447957029 Enamine

ethyl 3-bromo-4-fluoro-2-methylbenzoate

  • MDL number MFCD28397457
  • Cas Number : 1803604-00-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207461

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207461
Availability details for
Quantity
  • Availability
  • Add to Cart