ENA447957031 Enamine

4-(5-methylpyrrolidin-3-yl)morpholine

  • MDL number MFCD28397459
  • Cas Number : 1803587-99-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207465

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207465
Availability details for
Quantity
  • Availability
  • Add to Cart