ENA447957035 Enamine

sodium 6-amino-2,3-dihydro-1H-indene-5-sulfonate

  • MDL number MFCD28397463
  • Cas Number : 1803604-01-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Na+
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207502

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207502
Availability details for
Quantity
  • Availability
  • Add to Cart