ENA447957040 Enamine

2-(1-methanesulfonylcyclopropyl)propan-2-ol

  • MDL number MFCD28397469
  • Cas Number : 1803598-00-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207571

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207571
Availability details for
Quantity
  • Availability
  • Add to Cart