ENA447957041 Enamine

N-(4H-1,2,4-triazol-3-yl)pyridin-3-amine

  • MDL number MFCD28397470
  • Cas Number : 1803588-42-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207582

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207582
Availability details for
Quantity
  • Availability
  • Add to Cart