ENA447957042 Enamine

4-cyclopropyl-1H-imidazol-2-amine hydrochloride

  • MDL number MFCD28397471
  • Cas Number : 1803598-15-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207586

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207586
Availability details for
Quantity
  • Availability
  • Add to Cart