ENA447957044 Enamine

ethyl 5-methyl-1-(piperidin-4-yl)-1H-pyrazole-4-carboxylate hydrochloride

  • MDL number MFCD28397473
  • Cas Number : 1803593-01-2
  • Molecular Weight : 273.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-207600

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207600
Availability details for
Quantity
  • Availability
  • Add to Cart