ENA447957047 Enamine

1,4-diazepane-2-carboxylic acid dihydrobromide

  • MDL number MFCD28397476
  • Cas Number : 1803570-17-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HBr
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207614

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207614
Availability details for
Quantity
  • Availability
  • Add to Cart