ENA447957048 Enamine

3-hydroxy-1-methylpyridin-1-ium; tetrafluoroboranuide

  • MDL number MFCD28397477

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data BF4-
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207637

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207637
Availability details for
Quantity
  • Availability
  • Add to Cart