ENA447957050 Enamine

1-(2-methylphenyl)-1-(1H-pyrazol-1-yl)propan-2-amine dihydrochloride

  • MDL number MFCD28397479
  • Cas Number : 1803562-88-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207649

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207649
Availability details for
Quantity
  • Availability
  • Add to Cart