ENA447957051 Enamine

1-[(2S,4S)-4-fluoro-1-(5-methyl-1,3,4-oxadiazol-2-yl)pyrrolidin-2-yl]methanamine dihydrochloride

  • MDL number MFCD31566972
  • Cas Number : 1820574-42-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207651

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207651
Availability details for
Quantity
  • Availability
  • Add to Cart