ENA447957053 Enamine

2-[2-(aminomethyl)bicyclo[2.2.1]heptan-2-yl]acetic acid hydrochloride

  • MDL number MFCD28397482
  • Cas Number : 196618-71-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207685

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207685
Availability details for
Quantity
  • Availability
  • Add to Cart