ENA447957056 Enamine

ethyl 4-benzyl-3,5-dioxo-4-azatetracyclo[5.3.2.0^{2,6}.0^{8,10}]dodec-11-ene-9-carboxylate

  • Cas Number : 1803601-47-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207726

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207726
Availability details for
Quantity
  • Availability
  • Add to Cart