ENA447957058 Enamine

methyl[2-(3-phenyl-1,2,4-oxadiazol-5-yl)ethyl]amine hydrochloride

  • MDL number MFCD28397486
  • Cas Number : 1803605-77-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207747

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207747
Availability details for
Quantity
  • Availability
  • Add to Cart