ENA447957059 Enamine

2-(bromomethyl)morpholine hydrobromide

  • MDL number MFCD28397487
  • Cas Number : 1803588-05-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207788

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207788
Availability details for
Quantity
  • Availability
  • Add to Cart