ENA447957060 Enamine

3-chloro-2,2-dimethylpropan-1-amine hydrochloride

  • MDL number MFCD28397488
  • Cas Number : 1803588-46-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207792

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207792
Availability details for
Quantity
  • Availability
  • Add to Cart