ENA447957062 Enamine

ethyl 2,2-difluoro-3-(morpholin-4-yl)-3-oxopropanoate

  • MDL number MFCD28397490
  • Cas Number : 1357625-59-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207820

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207820
Availability details for
Quantity
  • Availability
  • Add to Cart