ENA447957063 Enamine

1-phenylpent-4-yn-2-amine hydrochloride

  • MDL number MFCD28397491
  • Cas Number : 1803593-13-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207826

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207826
Availability details for
Quantity
  • Availability
  • Add to Cart